p-Toluenethiol, 98%
p-Toluenethiol, 98%
p-Toluenethiol, 98%
Thermo Scientific Chemicals
p-Toluenethiol, 98%
CAS: 106-45-6 | C7H8S | 124.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA138715000
Price (MYR)
1,455.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,455.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Toluenethiol
Name Note98%
CAS106-45-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement