Thioanisole, 99%
Thioanisole, 99%
Thioanisole, 99%
Thermo Scientific Chemicals
Thioanisole, 99%
CAS: 100-68-5 | C7H8S | 124.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA138671000
Price (MYR)
414.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
414.00
EA
Add to cart
Specifications
Chemical Name or MaterialThioanisole
Name Note99%
CAS100-68-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.