Thianaphthene, 97%
Thianaphthene, 97%
Thianaphthene, 97%
Thermo Scientific Chemicals
Thianaphthene, 97%
CAS: 95-15-8 | C8H6S | 134.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA138620250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThianaphthene
CAS95-15-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.