Tetraphenylboron sodium, 99.5%
Tetraphenylboron sodium, 99.5%
Tetraphenylboron sodium, 99.5%
Tetraphenylboron sodium, 99.5%
Thermo Scientific Chemicals
Tetraphenylboron sodium, 99.5%
Tetraphenylboron sodium, 99.5%, C24H20BNa, CAS Number-143-66-8, borate 1-, tetraphenyl-, sodium, dotite kalibor, kalibor, kalignost, kariporu k, sodium tetraphenylborate, sodium tetraphenylborate 1-, sodium tetraphenylboride, sodium tetraphenylboron, tetraphenylboron sodium, 100g | CAS: 143-66-8 | C24H20BNa | 342.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA138521000100 gGlass bottle
Catalog number FSA138521000
Price (MYR)
1,693.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,693.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetraphenylboron sodium
Name Notep.a.
CAS143-66-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement