Tetramethylsilane, 99.9+%, NMR grade
Tetramethylsilane, 99.9+%, NMR grade
Tetramethylsilane, 99.9+%, NMR grade
Tetramethylsilane, 99.9+%, NMR grade
Thermo Scientific Chemicals
Tetramethylsilane, 99.9+%, NMR grade
CAS: 75-76-3 | C4H12Si | 88.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA138470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylsilane
Name Note99.9+%
CAS75-76-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Extremely flammable liquid and vapor.
View more
For NMR
RUO – Research Use Only