Tetramethylene sulfoxide, 97%
Tetramethylene sulfoxide, 97%
Tetramethylene sulfoxide, 97%
Tetramethylene sulfoxide, 97%
Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%
CAS: 1600-44-8 | C4H8OS | 104.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA138441000
Price (MYR)
2,384.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,384.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetramethylene sulfoxide
Name Note98+%
CAS1600-44-8
ColorColorless to Yellow
Density1.1500g/mL