Tetramethylene sulfone, 99+%
Tetramethylene sulfone, 99+%
Tetramethylene sulfone, 99+%
Tetramethylene sulfone, 99+%
Thermo Scientific Chemicals
Tetramethylene sulfone, 99+%
CAS: 126-33-0 | C4H8O2S | 120.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA138432500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylene sulfone
CAS126-33-0
Health Hazard 1GHS Signal Word: Warning
Melting Point27.4°C to 27.8°C
ColorYellow