Tetraethylthiuram Disulfide, 97%
Tetraethylthiuram Disulfide, 97%
Tetraethylthiuram Disulfide, 97%
Tetraethylthiuram Disulfide, 97%
Thermo Scientific Chemicals
Tetraethylthiuram Disulfide, 97%
CAS: 97-77-8 | C10H20N2S4 | 296.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA138125000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylthiuram disulfide
Name Note97%
CAS97-77-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning