Tetraethylenepentamine, technical
Tetraethylenepentamine, technical
Tetraethylenepentamine, technical
Tetraethylenepentamine, technical
Thermo Scientific Chemicals
Tetraethylenepentamine, technical
CAS: 112-57-2 | C8H23N5 | 189.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA138115000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylenepentamine
Name Notetech.
CAS112-57-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.