1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
Thermo Scientific Chemicals
1-Tetradecylamine, 98%
CAS: 2016-42-4 | C14H31N | 213.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA138100500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Tetradecylamine
CAS2016-42-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi