Tetrachloroethylene, 99%, extra pure
Tetrachloroethylene, 99%, extra pure
Tetrachloroethylene, 99%, extra pure
Tetrachloroethylene, 99%, extra pure
Thermo Scientific Chemicals
Tetrachloroethylene, 99%, extra pure
Tetrachloroethylene, 99%, C2Cl4, CAS Number-127-18-4, ankilostin, carbon dichloride, ethene, tetrachloro, ethylene tetrachloride, perc, perchlorethylene, perchloroethylene, tetrachlorethylene, tetrachloroethene, tetrachloroethylene, 1L, 0.003% max. (NaOH), 98.5% min. (GC), 01, 187 | CAS: 127-18-4 | C2Cl4 | 165.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA138015000
Price (MYR)
318.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
318.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetrachloroethylene
Alkalinity0.003% max. (NaOH)
CAS127-18-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Causes serious eye irritati