D(-)-Tartaric acid, 99%
D(-)-Tartaric acid, 99%
D(-)-Tartaric acid, 99%
D(-)-Tartaric acid, 99%
Thermo Scientific Chemicals
D(-)-Tartaric acid, 99%
CAS: 147-71-7 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA137862500
Price (MYR)
1,142.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,142.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(-)-Tartaric acid
Name Note99%
CAS147-71-7
Melting Point167°C to 171°C
Health Hazard 1Danger