(3-chloropropyl)trimethoxysilane, 98+%
(3-chloropropyl)trimethoxysilane, 98+%
(3-chloropropyl)trimethoxysilane, 98+%
(3-chloropropyl)trimethoxysilane, 98+%
Thermo Scientific Chemicals
(3-chloropropyl)trimethoxysilane, 98+%
CAS: 2530-87-2 | C6H15ClO3Si | 198.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA137285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(3-Chloropropyl)trimethoxysilane
CAS2530-87-2
CAS Min %98.0
Health Hazard 1Warning
CAS Max %100.0