Lidocaine, 97.5%
Lidocaine, 97.5%
Lidocaine, 97.5%
Thermo Scientific Chemicals
Lidocaine, 97.5%
CAS: 137-58-6 | C14H22N2O | 234.343 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number FSA137250100
Price (MYR)
262.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
262.00
EA
Add to cart
Specifications
Chemical Name or MaterialLidocaine
Melting Point66.0°C to 69.0°C
CAS137-58-6
CAS Min %96.5
ColorWhite to Yellow