2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
Thermo Scientific Chemicals
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%
CAS: 1131-01-7 | C10H12ClNO | 197.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA135440100
Price (MYR)
620.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
620.00
EA
Add to cart
Specifications
Chemical Name or Material2-Chloro-N-(2, 6-dimethylphenyl)acetamide
CAS1131-01-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0