Potassium hydroxide, ca. 85%, for analysis, pellets
Potassium hydroxide, ca. 85%, for analysis, pellets
Potassium hydroxide, ca. 85%, for analysis, pellets
Potassium hydroxide, ca. 85%, for analysis, pellets
Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA134062500
Price (MYR)
281.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
281.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydroxide
Name Noteca. 85 %
Boiling Point1320.0°C
CAS1310-58-3
Health Hazard 1Danger