4-Iodotoluene, 98%
4-Iodotoluene, 98%
4-Iodotoluene, 98%
Thermo Scientific Chemicals
4-Iodotoluene, 98%
CAS: 624-31-7 | C7H7I | 218.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass Bottle
Catalog number FSA133940500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodotoluene
Name Note98%
CAS624-31-7
CAS Min %97.5
CAS Max %100.0