N-Formylmorpholine, 99%
N-Formylmorpholine, 99%
N-Formylmorpholine, 99%
N-Formylmorpholine, 99%
Thermo Scientific Chemicals
N-Formylmorpholine, 99%
CAS: 4394-85-8 | C5H9NO2 | 115.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA133805000
Price (MYR)
1,098.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,098.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Formylmorpholine
Name Note99+%
CAS4394-85-8
CAS Min %99.0
Health Hazard 1HYGROSCOPIC