Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger | CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Change view
Quantity:
2.5 L
25 mL
1 L
Packaging:
Glass Bottle
Catalog number FSA133710025
Price (MYR)
3,315.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
3,315.00
EA
Add to cart
Chemical Identifiers
CAS7664-41-7, 67-56-1
Specifications
Chemical Name or MaterialAmmonia
ColorColorless to Yellow
Name Noteca. 7N Solution in Methanol
CAS7664-41-7
Health Hazard 1GHS Signal Word: Danger