Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Thermo Scientific Chemicals
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Plastic bottle
Catalog number FSA133700010
Price (MYR)
680.00
EA
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
680.00
EA
Add to cart
Chemical Identifiers
CAS7647-01-0, 67-63-0
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
ColorColorless to Yellow
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Highly flammable liquid and vapor.