3-Aminoisoxazole, 95%
3-Aminoisoxazole, 95%
3-Aminoisoxazole, 95%
3-Aminoisoxazole, 95%
Thermo Scientific Chemicals
3-Aminoisoxazole, 95%
CAS: 1750-42-1 | C3H4N2O | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA133530050
Price (MYR)
990.00
EA
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
990.00
EA
Add to cart
Specifications
Chemical Name or Material3-Aminoisoxazole
CAS1750-42-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0