Squaric Acid Dibutyl Ester, 97%
Squaric Acid Dibutyl Ester, 97%
Squaric Acid Dibutyl Ester, 97%
Squaric Acid Dibutyl Ester, 97%
Thermo Scientific Chemicals
Squaric Acid Dibutyl Ester, 97%
CAS: 2892-62-8 | C12H18O4 | 226.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA133170050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSquaric acid dibutyl ester
CAS2892-62-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.