3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
Thermo Scientific Chemicals
3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%
CAS: 134-96-3 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA132880250
Price (MYR)
399.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
399.00
BOT
Add to cart
Specifications
Chemical Name or Material3,5-Dimethoxy-4-hydroxybenzaldehyde
CAS134-96-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0