Styrene, 99%, extra pure, stabilized
Styrene, 99%, extra pure, stabilized
Styrene, 99%, extra pure, stabilized
Styrene, 99%, extra pure, stabilized
Thermo Scientific Chemicals
Styrene, 99%, extra pure, stabilized
CAS: 100-42-5 | C8H8 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA132790100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialStyrene
Name NoteStabilized
CAS100-42-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
May be fatal if swallowed and enters airways.<br/