Shikimic Acid, 98%
Shikimic Acid, 98%
Shikimic Acid, 98%
Thermo Scientific Chemicals
Shikimic Acid, 98%
CAS: 138-59-0 | C7H10O5 | 174.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA132700010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialShikimic acid
CAS138-59-0
Melting Point185.0°C to 191.0°C
ColorBeige to White or Gray
EINECS Number205-334-2