L-Serine, 99%
L-Serine, 99%
L-Serine, 99%
Thermo Scientific Chemicals
L-Serine, 99%
CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass Bottle
Catalog number FSA132661000
Price (MYR)
530.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
530.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL-Serine
Melting Point222.0°C
Ammonium200ppm max.
CAS56-45-1
Arsenic (As)1ppm max.