Semicarbazide hydrochloride, 99+%
Semicarbazide hydrochloride, 99+%
Semicarbazide hydrochloride, 99+%
Semicarbazide hydrochloride, 99+%
Thermo Scientific Chemicals
Semicarbazide hydrochloride, 99+%
CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA132645000
Price (MYR)
530.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
530.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSemicarbazide hydrochloride
CAS563-41-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio