Salicylhydrazide, 98+%
Salicylhydrazide, 98+%
Salicylhydrazide, 98+%
Salicylhydrazide, 98+%
Thermo Scientific Chemicals
Salicylhydrazide, 98+%
CAS: 936-02-7 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA132611000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSalicylhydrazide
Name Note98%
CAS936-02-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: