D(-)-Salicin, 99+%
D(-)-Salicin, 99+%
D(-)-Salicin, 99+%
Thermo Scientific Chemicals
D(-)-Salicin, 99+%
CAS: 138-52-3 | C13H18O7 | 286.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA132591000
Price (MYR)
2,468.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,468.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(-)-Salicin
Name Note99+%
CAS138-52-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement