Rutin, 97+%
Rutin, 97+%
Rutin, 97+%
Thermo Scientific Chemicals
Rutin, 97+%
CAS: 207671-50-9 | C27H30O16 | 610.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA132395000
Price (MYR)
2,434.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,434.00
BOT
Add to cart
Specifications
Chemical Name or MaterialRutin hydrate
Name Note97.0-102.0%
CAS207671-50-9
Health Hazard 1HYGROSCOPIC
Health Hazard 2LIGHT SENSITIVE