D(-)-Ribose, 99+%
D(-)-Ribose, 99+%
D(-)-Ribose, 99+%
Thermo Scientific Chemicals
D(-)-Ribose, 99+%
CAS: 50-69-1 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA132365000
Price (MYR)
2,973.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,973.00
EA
Add to cart
Specifications
Chemical Name or MaterialD(-)-Ribose
CAS50-69-1
Melting Point88.0°C to 92.0°C
ColorBeige to White or Yellow
EINECS Number200-059-4