Riboflavin, 98%
Riboflavin, 98%
Riboflavin, 98%
Thermo Scientific Chemicals
Riboflavin, 98%
CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA132355000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRiboflavin
CAS83-88-5
Absorbance0.31 to 0.33 (A373/A267), 0.36 to 0.39 (A444/A267)
Melting Point290.0°C
ColorYellow to Orange