Resorcinol, 98%
Resorcinol, 98%
Resorcinol, 98%
Thermo Scientific Chemicals
Resorcinol, 98%
CAS: 108-46-3 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA132292500
Price (MYR)
411.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
411.00
BOT
Add to cart
Specifications
Chemical Name or MaterialResorcinol
Name Note98%
CAS108-46-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.