Quinaldine, 95+%
Quinaldine, 95+%
Quinaldine, 95+%
Thermo Scientific Chemicals
Quinaldine, 95+%
CAS: 91-63-4 | C10H9N | 143.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA132172500250 mLGlass bottle
Catalog number FSA132172500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialQuinaldine
Name Note97%
CAS91-63-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement