(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%
(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%
(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%
(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%
Thermo Scientific Chemicals
(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%
CAS: 98-79-3 | C5H7NO3 | 129.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA132121000
Price (MYR)
467.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
467.00
BOT
Add to cart
Specifications
Chemical Name or Material(S)-(-)-2-Pyrrolidone-5-carboxylic acid
CAS98-79-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici