Pyrrolidine, 99+%
Pyrrolidine, 99+%
Pyrrolidine, 99+%
Thermo Scientific Chemicals
Pyrrolidine, 99+%
CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA132082500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPyrrolidine
Name Note99+%
CAS123-75-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.