Pyrrole-2-carboxaldehyde, 99%
Pyrrole-2-carboxaldehyde, 99%
Pyrrole-2-carboxaldehyde, 99%
Pyrrole-2-carboxaldehyde, 99%
Thermo Scientific Chemicals
Pyrrole-2-carboxaldehyde, 99%
CAS: 1003-29-8 | C5H5NO | 95.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA132060050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPyrrole-2-carboxaldehyde
Name Note99%
CAS1003-29-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: