3-(Hydroxymethyl)pyridine, 98%
3-(Hydroxymethyl)pyridine, 98%
3-(Hydroxymethyl)pyridine, 98%
3-(Hydroxymethyl)pyridine, 98%
Thermo Scientific Chemicals
3-(Hydroxymethyl)pyridine, 98%
CAS: 100-55-0 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA132000050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-(Hydroxymethyl)pyridine
CAS100-55-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0