2-Pyridylacetic acid hydrochloride, 99%
2-Pyridylacetic acid hydrochloride, 99%
2-Pyridylacetic acid hydrochloride, 99%
2-Pyridylacetic acid hydrochloride, 99%
Thermo Scientific Chemicals
2-Pyridylacetic acid hydrochloride, 99%
CAS: 16179-97-8 | C7H7NO2·HCl | 173.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA131950250
Price (MYR)
2,239.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,239.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Pyridylacetic acid hydrochloride
Name Note99%
CAS16179-97-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning