2,6-Pyridinedicarboxylic Acid, 99%
2,6-Pyridinedicarboxylic Acid, 99%
2,6-Pyridinedicarboxylic Acid, 99%
2,6-Pyridinedicarboxylic Acid, 99%
Thermo Scientific Chemicals
2,6-Pyridinedicarboxylic Acid, 99%
CAS: 499-83-2 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA131885000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Pyridinedicarboxylic acid
CAS499-83-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0