4-Pyridinecarboxaldehyde, 98%
4-Pyridinecarboxaldehyde, 98%
4-Pyridinecarboxaldehyde, 98%
4-Pyridinecarboxaldehyde, 98%
Thermo Scientific Chemicals
4-Pyridinecarboxaldehyde, 98%
CAS: 872-85-5 | C6H5NO | 107.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA131845000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Pyridinecarboxaldehyde
CAS872-85-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0