Pyridine, 99+%, extra pure
Pyridine, 99+%, extra pure
Pyridine, 99+%, extra pure
Pyridine, 99+%, extra pure
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA131780500
Price (MYR)
202.00
BOT
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
202.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
CAS110-86-1
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.