2-Pyrazinecarboxylic acid, 99%
2-Pyrazinecarboxylic acid, 99%
2-Pyrazinecarboxylic acid, 99%
2-Pyrazinecarboxylic acid, 99%
Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%
CAS: 98-97-5 | C5H4N2O2 | 124.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA131711000
Price (MYR)
818.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
818.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Pyrazinecarboxylic acid
Name Note99%
CAS98-97-5
CAS Min %98.5
CAS Max %100.0