Purine, 99%
Purine, 99%
Purine, 99%
Thermo Scientific Chemicals
Purine, 99%
CAS: 120-73-0 | C5H4N4 | 120.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA131670010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPurine
CAS120-73-0
Melting Point214°C to 217°C
ColorWhite to Beige
EINECS Number204-421-2