Propyl 4-hydroxybenzoate, 99+%
Propyl 4-hydroxybenzoate, 99+%
Propyl 4-hydroxybenzoate, 99+%
Propyl 4-hydroxybenzoate, 99+%
Thermo Scientific Chemicals
Propyl 4-hydroxybenzoate, 99+%
CAS: 94-13-3 | C10H12O3 | 180.2 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA131591000100 gPlastic bottle
Catalog number FSA131591000
Price (MYR)
275.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
275.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropyl 4-hydroxybenzoate
Name Note99+%
CAS94-13-3
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.