(1S)-(-)-alpha-Pinene, 98%
(1S)-(-)-alpha-Pinene, 98%
(1S)-(-)-alpha-Pinene, 98%
(1S)-(-)-alpha-Pinene, 98%
Thermo Scientific Chemicals
(1S)-(-)-alpha-Pinene, 98%
CAS: 7785-26-4 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA131275000
Price (MYR)
616.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
616.00
EA
Add to cart
Specifications
Chemical Name or Material(1S)-(-)-α-Pinene
CAS7785-26-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation or rash occurs: Get medical advice/attention.
IF ON SKIN (or hair): Take off immediately all contam