Phthalide, 99%
Phthalide, 99%
Phthalide, 99%
Thermo Scientific Chemicals
Phthalide, 99%
CAS: 87-41-2 | C8H6O2 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA131091000
Price (MYR)
308.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
308.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhthalide
Name Note99%
CAS87-41-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: