Phthalhydrazide, 99%
Phthalhydrazide, 99%
Phthalhydrazide, 99%
Phthalhydrazide, 99%
Thermo Scientific Chemicals
Phthalhydrazide, 99%
CAS: 1445-69-8 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA131060250
Price (MYR)
328.00
EA
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
328.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhthalhydrazide
Name Note99%
CAS1445-69-8
Melting Point>300°C