Phloroglucinol Dihydrate, 99%
Phloroglucinol Dihydrate, 99%
Phloroglucinol Dihydrate, 99%
Phloroglucinol Dihydrate, 99%
Thermo Scientific Chemicals
Phloroglucinol Dihydrate, 99%
CAS: 6099-90-7 | C6H6O3·2H2O | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass Bottle
Catalog number FSA131041000
Price (MYR)
475.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
475.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhloroglucinol dihydrate
CAS6099-90-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for