Specifications
Chemical Name or MaterialPhenyltrichlorosilane
Name Note95%
CAS98-13-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the respiratory tract.
View more